Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036545.1 | NP_001031622.1 | 6 - 7 | 757 | 78 | |
NM_001125502.1 | NP_001118974.1 | 8 - 9 | 1041 | 78 | ||
NM_001203783.2 | NP_001190712.1 | 8 - 9 | 1078 | 78 | ||
NM_001340823.1 | NP_001329480.1 | 8 - 9 | 1051 | 78 | ||
NM_001340824.1 | NP_001329479.1 | 8 - 9 | 1051 | 78 | ||
NM_001340825.1 | NP_001329481.1 | 8 - 9 | 1078 | 78 | ||
NM_117371.1 | NP_193038.1 | 6 - 7 | 757 | 78 | ||
NM_179039.4 | NP_849370.1 | 8 - 9 | 1163 | 78 | ||
GenBank mRNA | AF360324.1 | - | 8 - 9 | 1012 | 78 | |
AY056332.1 | - | 6 - 7 | 544 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |