Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YUC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340844.1 | NP_001328364.1 | 1 - 2 | 1133 | 75 | |
NM_001340845.1 | NP_001328363.1 | 2 - 3 | 948 | 75 | ||
NM_117399.4 | NP_193062.1 | 1 - 2 | 1133 | 75 | ||
GenBank mRNA | AY133691.1 | - | 1 - 2 | 679 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |