Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340852.1 | NP_001328594.1 | 7 - 8 | 1273 | 117 | |
NM_001340853.1 | NP_001328593.1 | 7 - 8 | 1242 | 117 | ||
NM_001340854.1 | NP_001328592.1 | 7 - 8 | 1212 | 117 | ||
NM_001340855.1 | NP_001328591.1 | 7 - 8 | 1701 | 117 | ||
NM_001340856.1 | NP_001328590.1 | 7 - 8 | 1576 | 117 | ||
NM_117408.4 | NP_567403.1 | 7 - 8 | 1273 | 117 | ||
NM_179042.1 | NP_849373.1 | 7 - 8 | 1212 | 117 | ||
GenBank mRNA | AY059783.1 | - | 7 - 8 | 1211 | 117 | |
AY117270.1 | - | 6 - 7 | 712 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |