Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117409.5 | NP_193071.2 | 12 - 13 | 1962 | 134 | |
NM_179209.2 | NP_849540.1 | 12 - 13 | 1810 | 134 | ||
GenBank mRNA | AY062505.1 | - | 12 - 13 | 1810 | 134 | |
BT008379.1 | - | 11 - 12 | 1680 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |