Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117409.5 | NP_193071.2 | 5 - 6 | 600 | 139 | |
NM_179209.2 | NP_849540.1 | 5 - 6 | 448 | 139 | ||
GenBank mRNA | AY062505.1 | - | 5 - 6 | 448 | 139 | |
BT008379.1 | - | 4 - 5 | 318 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |