Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117409.5 | NP_193071.2 | 9 - 10 | 1701 | 60 | |
NM_179209.2 | NP_849540.1 | 9 - 10 | 1549 | 60 | ||
GenBank mRNA | AY062505.1 | - | 9 - 10 | 1549 | 60 | |
BT008379.1 | - | 8 - 9 | 1419 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |