Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCSLA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340860.1 | NP_001329703.1 | 6 - 7 | 1190 | 79 | |
NM_001340861.1 | NP_001329704.1 | 5 - 6 | 1331 | 79 | ||
NM_117415.3 | NP_193077.2 | 6 - 7 | 1190 | 79 | ||
GenBank mRNA | KJ138761.1 | - | 6 - 7 | 973 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |