Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117416.3 | NP_567404.1 | 1 - 2 | 152 | 113 | |
GenBank mRNA | AF129478.1 | - | 1 - 2 | 84 | 113 | |
BT010589.1 | - | 1 - 2 | 75 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |