Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP83A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117451.4 | NP_193113.1 | 1 - 2 | 1030 | 78 | |
GenBank mRNA | AF428469.1 | - | 1 - 2 | 904 | 78 | |
AY075697.1 | - | 1 - 2 | 910 | 78 | ||
AY102146.1 | - | 1 - 2 | 883 | 78 | ||
D78599.1 | - | 1 - 2 | 905 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |