Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203793.2 | NP_001190722.1 | 1 | 478 | 154 | |
NM_001340886.1 | NP_001319924.1 | 1 | 478 | 154 | ||
NM_001340887.1 | NP_001328953.1 | 1 | 459 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |