Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRNEXO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117461.2 | NP_193123.2 | 1 - 2 | 295 | 93 | |
NM_202814.2 | NP_974543.1 | 1 - 2 | 310 | 93 | ||
GenBank mRNA | AF531179.1 | - | 1 - 2 | 300 | 93 | |
AJ404476.1 | - | 1 - 2 | 295 | 93 | ||
BT010908.1 | - | 1 - 2 | 260 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |