Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340905.1 | NP_001319933.1 | 6 - 7 | 2750 | 86 | |
NM_001340906.1 | NP_001329589.1 | 6 - 7 | 2693 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |