Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP97B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117600.6 | NP_193247.2 | 12 - 13 | 1498 | 136 | |
GenBank mRNA | AY062675.1 | - | 12 - 13 | 1406 | 136 | |
BT002582.1 | - | 12 - 13 | 1373 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |