Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP97B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117600.6 | NP_193247.2 | 6 - 7 | 1065 | 76 | |
GenBank mRNA | AY062675.1 | - | 6 - 7 | 973 | 76 | |
BT002582.1 | - | 6 - 7 | 940 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |