Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340998.1 | NP_001329368.1 | 6 - 7 | 1732 | 84 | |
NM_001340999.1 | NP_001329367.1 | 6 - 7 | 1905 | 84 | ||
NM_117609.3 | NP_567460.1 | 6 - 7 | 1507 | 84 | ||
GenBank mRNA | AF424573.1 | - | 6 - 7 | 1446 | 84 | |
AK226353.1 | - | 6 - 7 | 1449 | 84 | ||
AY142024.1 | - | 6 - 7 | 1338 | 84 | ||
M73467.1 | - | 1427 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |