Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341063.1 | NP_001329006.1 | 17 - 18 | 2372 | 100 | |
NM_001341064.1 | NP_001329005.1 | 17 - 18 | 2128 | 100 | ||
NM_001341065.1 | NP_001329007.1 | 15 - 16 | 1843 | 100 | ||
NM_117648.8 | NP_193292.3 | 18 - 19 | 2165 | 100 | ||
GenBank mRNA | EU915246.1 | - | 18 - 19 | 2165 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |