Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117682.3 | NP_193325.1 | 11 - 12 | 1045 | 72 | |
GenBank mRNA | BT029023.1 | - | 11 - 12 | 936 | 72 | |
X82825.1 | - | 958 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |