Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341097.1 | NP_001319958.1 | 16 - 17 | 2454 | 76 | |
NM_117729.8 | NP_193367.7 | 15 - 16 | 2406 | 76 | ||
GenBank mRNA | AF465831.1 | - | 15 - 16 | 2339 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |