Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341097.1 | NP_001319958.1 | 2 - 3 | 452 | 96 | |
NM_117729.8 | NP_193367.7 | 1 - 2 | 404 | 96 | ||
GenBank mRNA | AF465831.1 | - | 1 - 2 | 337 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |