Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117849.4 | NP_193476.1 | 1 - 2 | 495 | 76 | |
GenBank mRNA | AJ441252.1 | - | 1 - 2 | 151 | 76 | |
AK228587.1 | - | 1 - 2 | 333 | 76 | ||
BT005756.1 | - | 1 - 2 | 333 | 76 | ||
BT006112.1 | - | 1 - 2 | 151 | 76 | ||
U09332.1 | - | 1 - 2 | 298 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |