Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117880.3 | NP_193506.2 | 7 - 8 | 1428 | 68 | |
GenBank mRNA | AY133700.1 | - | 7 - 8 | 1428 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |