Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341241.1 | NP_001329809.1 | 3 - 4 | 2773 | 68 | |
NM_117886.3 | NP_567538.1 | 3 - 4 | 2620 | 68 | ||
GenBank mRNA | AK118703.1 | - | 3 - 4 | 2362 | 68 | |
BT005967.1 | - | 2 - 3 | 2261 | 68 | ||
KJ138747.1 | - | 2 - 3 | 2261 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |