Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117888.3 | NP_567540.2 | 2 - 3 | 1108 | 92 | |
GenBank mRNA | AK118728.1 | - | 2 - 3 | 1108 | 92 | |
AY550297.1 | - | 2 - 3 | 1044 | 92 | ||
BT005642.1 | - | 2 - 3 | 1044 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |