Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036581.1 | NP_001031658.1 | 1 - 2 | 179 | 102 | |
NM_117898.5 | NP_567543.1 | 1 - 2 | 342 | 102 | ||
GenBank mRNA | AK176287.1 | - | 1 - 2 | 179 | 102 | |
AK176618.1 | - | 1 - 2 | 181 | 102 | ||
AK221239.1 | - | 1 - 2 | 168 | 102 | ||
AK221371.1 | - | 1 - 2 | 168 | 102 | ||
AK229721.1 | - | 1 - 2 | 143 | 102 | ||
AK317033.1 | - | 1 - 2 | 174 | 102 | ||
BT000968.1 | - | 1 - 2 | 82 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |