Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036581.1 | NP_001031658.1 | 2 - 3 | 294 | 69 | |
NM_117898.5 | NP_567543.1 | 2 - 3 | 457 | 69 | ||
GenBank mRNA | AK176287.1 | - | 2 - 3 | 294 | 69 | |
AK176618.1 | - | 2 - 3 | 296 | 69 | ||
AK221239.1 | - | 2 - 3 | 283 | 69 | ||
AK221371.1 | - | 2 - 3 | 283 | 69 | ||
AK229721.1 | - | 2 - 3 | 258 | 69 | ||
AK317033.1 | - | 2 - 3 | 289 | 69 | ||
BT000968.1 | - | 2 - 3 | 197 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |