Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084936.2 | NP_001078405.1 | 2 - 3 | 434 | 69 | |
NM_001160778.2 | NP_001154250.1 | 2 - 3 | 470 | 69 | ||
NM_001203830.1 | NP_001190759.1 | 2 - 3 | 457 | 69 | ||
NM_001341252.1 | NP_001329342.1 | 3 - 4 | 448 | 69 | ||
NM_001341253.1 | NP_001329343.1 | 3 - 4 | 428 | 69 | ||
NM_001341254.1 | NP_001329339.1 | 2 - 3 | 709 | 69 | ||
NM_001341255.1 | NP_001329340.1 | 2 - 3 | 689 | 69 | ||
NM_001341256.1 | NP_001329341.1 | 2 - 3 | 288 | 69 | ||
NM_117912.4 | NP_567548.1 | 2 - 3 | 470 | 69 | ||
NM_179072.3 | NP_849403.1 | 2 - 3 | 442 | 69 | ||
NM_179073.4 | NP_849404.2 | 2 - 3 | 410 | 69 | ||
GenBank mRNA | AB041531.1 | - | 1 - 2 | 160 | 69 | |
AF385725.1 | - | 2 - 3 | 294 | 69 | ||
AY143963.1 | - | 1 - 2 | 150 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |