Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRANS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117937.5 | NP_193561.4 | 1 - 2 | 335 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |