Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341273.1 | NP_001329801.1 | 2 - 3 | 1016 | 92 | |
NM_117939.5 | NP_193563.3 | 2 - 3 | 392 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |