Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341273.1 | NP_001329801.1 | 8 - 9 | 1920 | 107 | |
NM_117939.5 | NP_193563.3 | 8 - 9 | 1296 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |