Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341301.1 | NP_001319986.1 | 1 | 733 | 94 | |
GenBank mRNA | BT010856.1 | - | 1 | 636 | 94 | |
BT014677.1 | - | 1 | 540 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |