Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117980.5 | NP_193599.3 | 6 - 7 | 1298 | 108 | |
GenBank mRNA | FJ708745.1 | - | 5 - 6 | 583 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |