Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147892.4 | NP_680197.2 | 12 - 13 | 1535 | 67 | |
GenBank mRNA | BT022108.1 | - | 12 - 13 | 1456 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |