Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105215.3 | NP_564850.1 | 8 - 9 | 1103 | 75 | |
GenBank mRNA | AF361573.1 | - | 8 - 9 | 1103 | 75 | |
AY093985.1 | - | 8 - 9 | 910 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |