Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105234.3 | NP_176738.1 | 3 - 4 | 1540 | 137 | |
GenBank mRNA | AK229402.1 | - | 3 - 4 | 1397 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |