Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105236.2 | NP_564856.1 | 5 - 6 | 1206 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |