Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105327.2 | NP_176829.1 | 1 - 2 | 311 | 155 | |
GenBank mRNA | AY071850.1 | - | 1 - 2 | 250 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |