Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198404.1 | NP_001185333.1 | 5 - 6 | 1551 | 158 | |
NM_001334262.1 | NP_001321449.1 | 5 - 6 | 1801 | 158 | ||
NM_105368.3 | NP_176869.2 | 5 - 6 | 1727 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |