Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105405.1 | NP_564896.1 | 20 - 21 | 1750 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |