Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334301.1 | NP_001319333.1 | 2 - 3 | 240 | 99 | |
NM_001334302.1 | NP_001320390.1 | 2 - 3 | 291 | 99 | ||
NM_001334303.1 | NP_001320389.1 | 2 - 3 | 272 | 99 | ||
NM_001334304.1 | NP_001320393.1 | 2 - 3 | 268 | 99 | ||
NM_001334305.1 | NP_001320391.1 | 2 - 3 | 267 | 99 | ||
NM_001334306.1 | NP_001320392.1 | 2 - 3 | 281 | 99 | ||
NM_105417.4 | NP_176917.2 | 2 - 3 | 240 | 99 | ||
GenBank mRNA | AB114052.1 | - | 1 - 2 | 108 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |