Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198420.1 | NP_001185349.1 | 4 - 5 | 695 | 84 | |
NM_001334339.1 | NP_001320607.1 | 4 - 5 | 673 | 84 | ||
NM_105471.4 | NP_176968.1 | 4 - 5 | 698 | 84 | ||
GenBank mRNA | AF370221.1 | - | 4 - 5 | 538 | 84 | |
AK226564.1 | - | 515 | 84 | |||
AY099552.1 | - | 4 - 5 | 530 | 84 | ||
AY113871.1 | - | 4 - 5 | 473 | 84 | ||
BT001237.1 | - | 4 - 5 | 473 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |