Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334339.1 | NP_001320607.1 | 8 - 9 | 993 | 89 | |
NM_105471.4 | NP_176968.1 | 8 - 9 | 1018 | 89 | ||
GenBank mRNA | AF370221.1 | - | 8 - 9 | 858 | 89 | |
AK226564.1 | - | 835 | 89 | |||
AY099552.1 | - | 8 - 9 | 850 | 89 | ||
AY113871.1 | - | 8 - 9 | 793 | 89 | ||
BT001237.1 | - | 8 - 9 | 793 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |