Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105479.2 | NP_564920.1 | 5 - 6 | 862 | 109 | |
GenBank mRNA | DQ446409.1 | - | 5 - 6 | 741 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |