Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334346.1 | NP_001322211.1 | 6 - 7 | 1067 | 117 | |
NM_105480.3 | NP_564921.1 | 6 - 7 | 1076 | 117 | ||
GenBank mRNA | AF216524.1 | - | 6 - 7 | 901 | 117 | |
AK229148.1 | - | 6 - 7 | 958 | 117 | ||
AY084533.1 | - | 6 - 7 | 972 | 117 | ||
BT001920.1 | - | 6 - 7 | 851 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |