Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334365.1 | NP_001323149.1 | 1 - 2 | 124 | 82 | |
NM_105519.4 | NP_177015.3 | 2 - 3 | 359 | 82 | ||
GenBank mRNA | AY465924.1 | - | 2 - 3 | 329 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |