Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105527.5 | NP_177023.1 | 2 - 3 | 1116 | 126 | |
GenBank mRNA | AY057482.1 | - | 2 - 3 | 977 | 126 | |
BT002675.1 | - | 2 - 3 | 879 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Arabidopsis thaliana Probe spans exons |