Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105548.5 | NP_177043.2 | 1 - 2 | 201 | 85 | |
GenBank mRNA | AJ532903.1 | - | 1 - 2 | 158 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |