Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105557.6 | NP_564946.1 | 3 - 4 | 783 | 70 | |
GenBank mRNA | BT030352.1 | - | 3 - 4 | 594 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |