Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334400.1 | NP_001320362.1 | 4 - 5 | 682 | 87 | |
NM_001334401.1 | NP_001320361.1 | 4 - 5 | 436 | 87 | ||
NM_105581.3 | NP_177074.1 | 4 - 5 | 682 | 87 | ||
GenBank mRNA | BT004113.1 | - | 4 - 5 | 577 | 87 | |
BT004951.1 | - | 4 - 5 | 435 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |