Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105666.2 | NP_177155.1 | 1 | 239 | 81 | |
GenBank mRNA | DQ446415.1 | - | 1 | 239 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |