Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105699.4 | NP_177188.1 | 1 - 2 | 422 | 138 | |
GenBank mRNA | AF375409.1 | - | 1 - 2 | 239 | 138 | |
AJ251297.1 | - | 1 - 2 | 211 | 138 | ||
BT020360.1 | - | 1 - 2 | 179 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |